Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BNP Antibody (SR1505), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | BNP |
---|---|
Clone | SR1505 |
Dilution | Western Blot 1:1000-1:5000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
BNP Monoclonal antibody specifically detects BNP in Human samples. It is validated for Western BlotSpecifications
BNP | |
Western Blot 1:1000-1:5000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein | |
A synthesized peptide derived from human BNP (Uniprot #: P16860) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1505 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
4879 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title