Learn More
Invitrogen™ BOB-1 Monoclonal Antibody (SP92)

Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil tissue.
Specifications
Specifications
| Antigen | BOB-1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | SP92 |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.1% sodium azide; pH 7.6 |
| Gene | POU2AF1 |
| Gene Accession No. | Q16633 |
| Gene Alias | B-cell-specific coactivator OBF-1; BOB.1; Bob1; BOB-1; OBF.1; OBF1; Obf-1; OCAB; OCA-B; OCT-binding factor 1; POU class 2 associating factor 1; POU domain class 2-associating factor 1; POU domain, class 2, associating factor 1; Pou2af1 |
| Gene Symbols | POU2AF1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.