Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOB1 Antibody (BOB1/2423), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB391529 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391529 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391529 Supplier Novus Biologicals Supplier No. NBP308666AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BOB1 Monoclonal antibody specifically detects BOB1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen BOB1
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone BOB1/2423
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias B-cell-specific coactivator OBF-1, BOB-1, OBF-1, OBF1BOB1, OCAB, OCA-B, OCT-binding factor 1, POU class 2 associating factor 1, POU domain class 2, associating factor 1, POU domain class 2-associating factor 1, POU domain, class 2, associating factor 1
Host Species Mouse
Immunogen Recombinant fragment (around aa 148-255) of human BOB1 protein (exact sequence is proprietary) (Uniprot: Q16633)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5450
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.