Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexakis(1H,1H,omegaH-perfluoroalkoxy)phosphazene, mass spectrometry standard, Thermo Scientific™

Catalog No. AC449170010
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 105809-15-2
Molecular Formula C6H18N3P3
Molecular Weight (g/mol) 2120
InChI Key DEBZEVJNWCNATM-MZWXYZOWSA-N
PubChem CID 44119860
IUPAC Name 2,2,4,4,6,6-hexakis(deuteriomethyl)-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
SMILES CP1(=NP(=NP(=N1)(C)C)(C)C)C

Specifications

Density 1.8000g/mL
Packaging Glass bottle
Refractive Index 1.3490 to 1.3570
Quantity 1 g
Specific Gravity 1.8
Synonym ultramark1621, ultramark 1621, hexa 2 h? methyl-1,3,5,2??,4??,6??-triazatriphosphinine
Formula Weight 2120
Chemical Name or Material Hexakis(1H, 1H, ωH-perfluoroalkoxy)phosphazene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.