Learn More
Description
Specifications
Specifications
| Antigen | BOLA2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | BolA Homolog 2B (E. Coli), BOLA2, BOLA2A, BOLA2B, BolA-Like 2B, BolA-Like 2B (E. Coli), BolA-Like Protein 2, BolA-Like Protein 2 Member B, BolA-Like Protein 2B |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the n terminal region of mouse BOLA2 (NP_780312). Peptide sequence MELSADYLREKLRQDLEAEHVEVEDTTLNRCATSFRVLVVSAKFEGKPLL |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
