Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bone marrow stromal cell antigen 1/CD157 Antibody (SY11B5), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N237711P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N237711P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N237711P594 Supplier Novus Biologicals Supplier No. NBP237711PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bone marrow stromal cell antigen 1/CD157 Monoclonal specifically detects Bone marrow stromal cell antigen 1/CD157 in Human, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen Bone marrow stromal cell antigen 1/CD157
Applications Flow Cytometry
Classification Monoclonal
Clone SY11B5
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ADP-ribosyl cyclase 2, Bone marrow stromal antigen 1, bone marrow stromal cell antigen 1, BST-1, cADPr hydrolase 2, CD157, CD157 antigen, Cyclic ADP-ribose hydrolase 2, EC 3.2.2.5, NAD(+) nucleosidase
Gene Symbols BST1
Host Species Mouse
Immunogen Human CD157.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 683.0
Test Specificity The mouse monoclonal antibody SY11B5 recognizes CD157, an approximately 45 kDa GPI-anchored protein expressed mainly on monocytes, macrophages, granulocytes and bone marrow stromal cells.
Target Species Human, Primate
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.