Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ BOP-JF549
Click to view available options
Quantity:
100 μg
Description
BOP-JF549 is a fluorescent dual α9β1/α4β1 integrin inhibitor. Comprises BOP (Cat. No. 6047) conjugated to Janelia Fluor™ 549 (Cat. No. 6147). Fluorogenic: fluoresces only once bound to integrins, enabling hassle-free no-wash experiments. Bright and photostable, enabling live cell tracking of integrin receptors over long time-course experiments. Excitation maximum = 549 nm; emission maximum = 571 nm.
Specifications
Specifications
Chemical Name or Material | 2-(3-(Azetidin-1-ium-1-ylidene)-6-(azetidin-1-yl)-3 H-xanthen-9-yl)-4-(((1-((3 R,5 S)-5-((-1-carboxy-2-(4-((pyrrolidine-1-carbonyl)oxy)phenyl)ethyl)carbamoyl)-1-(phenylsulfonyl)pyrrolidin-3-yl)-1 H-1,2,3-triazol-4-yl)methyl)carbamoyl)benzoate |
Quantity | 100 μg |
Target | Fluorescent Integrins Probes |
Molecular Formula | C55H53N9O11S |
Purity | 0.85 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction