Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOP1 Antibody (T7-P1B7*A8), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP250124AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250124AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250124AF Supplier Novus Biologicals Supplier No. NBP250124AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BOP1 Monoclonal specifically detects BOP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen BOP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone T7-P1B7*A8
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias block of proliferation 1, KIAA0124Block of proliferation 1 protein, ribosome biogenesis protein BOP1
Gene Symbols BOP1
Host Species Mouse
Immunogen Peptide Sequence - KGHVLTRDLG
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 23246
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.