Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Borate Buffer, pH 9.5, for Ammonia and Organic Nitrogen Analysis, Reagents
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
Specifications
Specifications
Chemical Name or Material | Buffer |
CAS | 7732-18-5 |
Color | Colorless |
pH | 9.5 |
Physical Form | Liquid |
Quantity | 4 L |
Packaging | Cubitainer |
Sterility | Non-sterile |
Solubility Information | Completely miscible |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction