Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bordetella Pertussis Toxin Antibody (1280/204), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB10064514Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064514Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064514Y Supplier Novus Biologicals Supplier No. NB10064514AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bordetella Pertussis Toxin Monoclonal specifically detects Bordetella Pertussis Toxin in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Bordetella Pertussis Toxin
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1280/204
Conjugate Alexa Fluor 647
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias pertussis toxin, pertussis toxin A protomer
Host Species Mouse
Immunogen Native
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64514 is reactive with Bordetella pertussis toxin in a simple ELISA. Reactivities with other organisms have not yet been determined.
Reconstitution Protein A purified
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.