Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borealin Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP122832B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122832B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122832B Supplier Novus Biologicals Supplier No. NBP122832B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Borealin Polyclonal specifically detects Borealin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Borealin
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q53HL2
Gene Alias BOR, BOREALIN, cell division cycle associated 8, Cell division cycle-associated protein 8, Dasra B, DasraB, Dasra-B, FLJ10468, FLJ12042, hDasra-B, MESRGP, PESCRG3, Pluripotent embryonic stem cell-related gene 3 protein
Gene Symbols CDCA8
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Borealin (within residues 200-250). [Swiss-Prot# Q53HL2]
Molecular Weight of Antigen 38 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 55143
Test Specificity Human. Immunogen sequence has 92% identity with mouse and rat proteins.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.