Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borealin Antibody (OTI4G3), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288471 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288471 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288471 Supplier Novus Biologicals Supplier No. NBP271988APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Borealin Monoclonal antibody specifically detects Borealin in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Borealin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4G3
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias BOR, BOREALIN, cell division cycle associated 8, Cell division cycle-associated protein 8, Dasra B, dasra-B, DasraB, FLJ10468, FLJ12042, hDasra-B, MESRGP, PESCRG3, Pluripotent embryonic stem cell-related gene 3 protein
Host Species Mouse
Immunogen Full length human recombinant protein of human CDCA8 (NP_060571) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 55143
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.