Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Granular, NF, EP, BP, JP, 99.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601050 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-050 12 kg Poly Pail
18-601-047 125 g Poly Bottle
18-601-048 500 g Poly Bottle
18-601-049 2.5 kg Poly Pail
18-601-051 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-050 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. BO12012KG
Only null left
Add to Cart
Add to Cart

BO120, 10043-35-3, H3BO3

Spectrum™ Chemical Boric Acid, Granular, NF, EP, BP, JP is used as an antimicrobial preservative, a buffering agent and as a tonicity adjusting agent. Spectrum Chemical NF, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8-4.8
CAS Max % 100%
Packaging Poly Pail
Quantity 12 kg
Percent Purity 99.5 to 100.5%
Grade Multi-Compendial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.