Learn More
Boric acid, Puratronic™, 99.9995% (metals basis)
Description
It is used as a primary or adjunct buffer in swimming pools, flame retardant, neutron absorber in nuclear power plants, colorant to make flame green, preservative in urine samples, welding flux in combination with borax, or precursor to other chemical compounds. It is used to prevent/remove existing wet and dry rot in timbers, treat wood against fungal/insect attack, prevent athlete’s foot, lubricate carom boards, and reduce surface oxidation on metals at high temperatures. It can be used to produce lubricant for ceramic or metal surfaces, textile fiberglass, induction furnace linings and ceramics, Silly Putty, and glass LCD flat panel displays. It can prevent the high temperature reaction of aluminum with nitrates in pyrotechniques, and neutralize active hydrofluoric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∽185°C (decomposition) |
| Density | 1.43515 |
| Flash Point | None |
| Assay Percent Range | 99.9995% (metals basis) |
| Quantity | 25 g |
| Physical Form | Powder |
| Linear Formula | H3BO3 |
| Beilstein | 1697939 |
| Merck Index | 14,1336 |
| Solubility Information | Soluble in water- solubility increases rapidly with temperature. Soluble in methanol and ethanol. Very slightly soluble in acetone. |
| Show More |
Safety and Handling
| Hazard Category | H360FD |
| Hazard Statement | GHS H Statement H360 May damage fertility or the unborn child. |
| Precautionary Statement | P201-P202-P260-P281-P308+P313-P501c |
| EINECSNumber | 233-139-2 |
| RTECSNumber | ED4550000 |
| TSCA | Yes |
| storageNote1 | Hygroscopic |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.