Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid Solution with Indicator, Ricca Chemical
SDP

Catalog No. SA950010 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA9500-10 10 L Cubitainer
SA95001 1 L Natural Poly Bottle
SA95004 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SA9500-10
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 10043-35-3 , 64-17-5 , 67-63-0 , 67-56-1 , 76-60-8
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

CAS Min % 3.76, 94.0, 0.2, 0.01, 0.1, 1.0E-4
CAS Max % 3.92, 98.0, 0.3, 0.02, 0.2, 1.0E-4
Packaging Cubitainer
Quantity 10 L
Percent Purity 4% W/V
Grade Laboratory
Chemical Name or Material Boric Acid Solution with Indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.