Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BORIS Antibody (20B11) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB413837 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB413837 0.025 mL
NBP252405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB413837 Supplier Novus Biologicals Supplier No. NBP2524050.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

BORIS Monoclonal specifically detects BORIS in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen BORIS
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 20B11
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias BORISHMGB1L1, BORIS-like protein, Brother of the regulator of imprinted sites, Cancer/testis antigen 27, CCCTC-binding factor, CCCTC-binding factor (zinc finger protein)-like, CT27MGC163358, CTCF paralog, CTCF-like protein, CTCF-T, dJ579F20.2, HMG-1L1, MGC169105, MGC169106, putative high mobility group protein 1-like 1, putative high mobility group protein B1-like 1, transcriptional repressor CTCFL, Zinc finger protein CTCF-T
Gene Symbols CTCFL
Host Species Mouse
Immunogen Synthetic peptide within the BORIS C-terminal domain (aa 614-648) (CG)GEMFPVACRETTARVKEE (NB - the first two aa do not belong to BORIS)
Molecular Weight of Antigen 83 kDa
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140690
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.