Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BORIS Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB119939 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119939 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119939 Supplier Novus Biologicals Supplier No. NB1002028FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BORIS Polyclonal specifically detects BORIS in Human samples. It is validated for ELISA, Western Blot.

Specifications

Antigen BORIS
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias BORISHMGB1L1, BORIS-like protein, Brother of the regulator of imprinted sites, Cancer/testis antigen 27, CCCTC-binding factor, CCCTC-binding factor (zinc finger protein)-like, CT27MGC163358, CTCF paralog, CTCF-like protein, CTCF-T, dJ579F20.2, HMG-1L1, MGC169105, MGC169106, putative high mobility group protein 1-like 1, putative high mobility group protein B1-like 1, transcriptional repressor CTCFL, Zinc finger protein CTCF-T
Host Species Rabbit
Immunogen This affinity purified BORIS Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to N-Terminal region near aa 1-30 of human BORIS protein. (Uniprot: Q8NI51)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140690
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.