Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron Powder, amorphous and crystalline, -325 mesh, 98% (Metals basis), Thermo Scientific Chemicals

Catalog No. AA1133730
Click to view available options
Quantity:
10 g
50 g
250 g

Catalog No. AA1133730

Supplier: Thermo Scientific Chemicals 1133730

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-42-8
Molecular Formula B
Molecular Weight (g/mol) 10.81
MDL Number MFCD00134034 MFCD00151272
InChI Key ZOXJGFHDIHLPTG-UHFFFAOYSA-N
Synonym boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
PubChem CID 5462311
ChEBI CHEBI:27560
IUPAC Name boron
SMILES [B]

Specifications

Color Brown
Physical Form Powder, Amorphous and Crystalline
Quantity 250 g
Assay Percent Range 98% (Metals basis)
Ignition Point 700°C (1292°F)
Odor Odorless
Appearance Brown powder
Chemical Name or Material Boron
Hazard Category H228-H302-H332-H335; Flammable solid. Harmful if swallowed. Harmful if inhaled. May cause respiratory irritation.
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 231-151-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.