Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron trifluoride etherate, approx. 48% BF3, ACROS Organics™

Click to view available options
Quantity:
25g
100g
1kg
2.5kg
Packaging:
Glass bottle
109-63-7
C4H10BF3O
141.93
MFCD00013194
KZMGYPLQYOPHEL-UHFFFAOYSA-N
Boron trifluoride ethyl ether
8000
ethoxyethane;trifluoroborane
B(F)(F)F.CCOCC
109-63-7
C4H10BF3O
141.93
MFCD00013194
KZMGYPLQYOPHEL-UHFFFAOYSA-N
Boron trifluoride ethyl ether
8000
ethoxyethane;trifluoroborane
B(F)(F)F.CCOCC
Lewis acid
Color Colorless to Straw-Yellow
Refractive Index 1.3429 to 1.3449
Packaging Glass bottle
Viscosity 1.89 mPa.s (20°C)
Melting Point -60.0°C
Flash Point 58°C
Quantity 100g
Density 1.1200g/mL
Boiling Point 126.0°C
Specific Gravity 1.12
Linear Formula (C2H5)2O·BF3
Fieser 01,70; 02,35; 03,33; 04,44; 05,52; 06,65; 07,31; 08,51; 09,64; 10,52; 11,72; 12,66; 13,3; 14,54; 15,45; 16,44; 17,52
Merck Index 15,1352
Solubility Information Solubility in water: reacts. Other solubilities: decomposes in alcohol,soluble in ether
Formula Weight 141.93
Physical Form Liquid, Fuming In Moist Air
Percent Purity approx. 48% BF3
Chemical Name or Material Boron trifluoride etherate, 48% BF3
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Danger

EINECSNumber : 203-689-8

TSCA : TSCA

Recommended Storage : Product darkens on storage