Learn More
Cytiva Whatman™ Borosilicate Glass Frits
Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
Whatman GV050 Series Glass Vacuum Filtration System
Description
- Material: Borosilicate Glass
- For Use With (Application): Filtration (aqueous and organic solvents) and biological analysis
Specifications
Specifications
Disposable | Reusable |
For Use With (Equipment) | Whatman GV050 Series Glass Vacuum Filtration System |
Material | Borosilicate Glass |
Sterility | Non-sterile |
For Use With (Application) | Vacuum Filtration |
Model | GV050/0/03 |
Quantity | 1 Ea. |
Type | Glass Frit Filter |
For internal research use only. Products are not intended for diagnostic use or resale.