Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borrelia Burgdorferi Antibody (BGN/F44/42), Alexa Fluor™ 350
SDP

Catalog No. N1066394A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1066394A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1066394A30 Supplier Novus Biologicals Supplier No. NB10066394AF350

Mouse Monoclonal Antibody

Borrelia Burgdorferi Monoclonal specifically detects Borrelia Burgdorferi in Bacteria samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Borrelia Burgdorferi
Applications Western Blot, ELISA
Classification Monoclonal
Clone BGN/F44/42
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias B burgdorferi, B. burgdorferi, B.burgdorferi, Lyme disease
Host Species Mouse
Immunogen Native
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity NB100-66394 recognizes Borrelia burgdorferi, a gram-negative spirochete that is a causative agent of lyme disease in humans.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.