Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borrelia Burgdorferi Antibody (BGN/F44/42) - BSA Free
SDP

Catalog No. NB017768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017768 0.025 mg
NB10066394 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017768 Supplier Novus Biologicals Supplier No. NB100663940.025MG

Mouse Monoclonal Antibody

Borrelia Burgdorferi Monoclonal specifically detects Borrelia Burgdorferi in Bacteria samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Borrelia Burgdorferi
Applications ELISA, Western Blot
Classification Monoclonal
Clone BGN/F44/42
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation PBS with 0.09% Sodium Azide
Gene Alias B burgdorferi, B. burgdorferi, B.burgdorferi, Lyme disease
Host Species Mouse
Immunogen Native
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-66394 recognizes Borrelia burgdorferi, a gram-negative spirochete that is a causative agent of lyme disease in humans.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.