Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borrelia Burgdorferi Flagellar Antigen Antibody (7G11/6802), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB11059982Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11059982Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11059982Z Supplier Novus Biologicals Supplier No. NB11059982AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Borrelia Burgdorferi Flagellar Antigen Monoclonal specifically detects Borrelia Burgdorferi Flagellar Antigen in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Borrelia Burgdorferi Flagellar Antigen
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 7G11/6802
Conjugate Alexa Fluor 700
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias B burgdorferi, B. burgdorferi, B.burgdorferi, Lyme disease
Host Species Mouse
Immunogen Native
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with a flagellar antigen.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.