Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borrelia burgdorferi garinii Monoclonal Antibody (Bg14), Invitrogen™

Catalog No. PIMA121681
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA121681 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA121681 Supplier Thermo Scientific Supplier No. MA121681

Mouse Monoclonal Antibody

MA1-21681 detects Borrelia burgdorferi garinii in Bacteria samples. MA1-21681 has been successfully used in ELISA, Immunofluorescence and Western Blot procedures. MA1-21681 immunogen corresponds to Bacterial lysates of Borrelia burgdorferi garinii..

Borrelia burgdorferi is a spirochete and the cause of Lyme disease, a tick transmitted illness of humans and animals. B. burgdorferi may persist in humans and animals for months or years following initial infection, despite a robust humoral immune response. B. burgdorferi resembles other spirochetes in that it is a highly specialized, motile, two-membrane, spiral shaped bacteria which lives primarily as an extracellular pathogen. B. burgdorferi has an unusual genome compared with other eubacteria which includes a linear chromosome approximately one megabase in size and numerous linear and circular plasmids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Borrelia burgdorferi garinii
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone Bg14
Concentration 2.8 mg/mL
Conjugate Unconjugated
Gene Alias B. burgdorferi; Lyme disease; Borrelia
Host Species Mouse
Immunogen Bacterial lysates of Borrelia burgdorferi garinii.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4° C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.