Learn More
United Scientific™ Bottle Top Dispenser with Bottle

Shop All United Scientific Supplies, Inc Products


Description
United brand bottle top dispensers offer high accuracy at economical prices.
- Each dispenser is individually calibrated and certified in an ISO 17025 accredited laboratory in accordance with ISO 8655 standards
- A set of adapters to fit most laboratory reagent bottles are provided
- All wetted parts are made from PTFE and borosilicate glass for maximum chemical resistance
- These easy-to-use instruments are suitable for all laboratory applications, except HF
- Special precautions must be taken for use, cleaning, and storage when working with concentrated fuming acids
- A factory calibration certificate is provided with each bottle top dispenser
- Dispensers are easy to disassemble and can be recalibrated in the lab
- Fully autoclavable at 121°C, 15 psi for 10 to 15 minutes
Specifications
Specifications
| Accuracy | 0.005 |
| Product Type | Analog Bottletop Dispenser with Adapters |
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO 8655 |
| Color | White |
| Channels | 1 |
| Material | PTFE, Borosilicate Glass |
| Volume (Metric) | 0.5 to 5 mL |
| Display | Analog |
| For Use With (Application) | Dispensing Liquids |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.