Learn More
PIP™ Bouton™ Bullseye™ Rimless Safety Glasses
Trendy design that hits the mark with a lens treatment to fit your application.
Catalog Number | Mfr. No. | Lens Coating | Lens Tint | Frame Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Coating | Lens Tint | Frame Color | Price | Quantity & Availability | |||
19-153-341
|
PIP™
250310021 |
Scratch-resistant | Gray | Gray | N/A | N/A | N/A | ||
19-153-344
|
PIP™
250310010 |
Anti-reflective, Scratch-resistant | Clear | Clear | N/A | N/A | N/A | ||
19-153-342
|
PIP™
250310023 |
Scratch-resistant | Light Blue | Light Blue | N/A | N/A | N/A | ||
19-153-347
|
PIP™
250310031 |
Scratch-resistant | Gradient | Clear | N/A | N/A | N/A | ||
Description
- Lightweight, rimless design offers optimal protection
- Flexible bayonet style temples provide a secure fit
- Molded nose bridge design to fit the vast majority of users
- Clear Lens - clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
- Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
- Anti-Fog Coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Specifications
Safety glasses | |
Polycarbonate |