Learn More
PIP™ Bouton™ Commander™ Semi-Rimless Safety Glasses
Unique styling that will command your attention with fit, comfort and a variety of technical lens treatments.
$177.10 - $425.53
Specifications
Product Type Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
View More Specs
Products 10
Catalog Number Mfr. No. Frame Color Lens Coating Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Frame Color Lens Coating Lens Tint Price Quantity & Availability  
19-153-369
View Documents
PIP™
250330020
Gloss Black Anti-fog, Scratch-resistant Clear
Dozen of 12 for $177.10
Case of 12 DZ for $2,125.19
Only null left
Add to Cart
19-153-370
View Documents
PIP™
250330021
Gloss Black Anti-fog, Scratch-resistant Gray
Dozen of 12 for $262.21
Case of 12 DZ for $3,146.51
Only null left
Add to Cart
19-153-372
View Documents
PIP™
250330226
Gloss Black Anti-fog, Scratch-resistant Blue, Indoor/Outdoor
Dozen of 12 for $364.47
Case of 12 DZ for $4,373.66
Only null left
Add to Cart
19-153-373
View Documents
PIP™
250330041
Gloss Black Scratch-resistant Gray
Pack of 6 for $425.53
Case of 12 PK for $5,106.36
Only null left
Add to Cart
19-153-376
View Documents
PIP™
250331010
Gloss Bronze Scratch-resistant Clear
Dozen of 12 for $367.44
Case of 12 DZ for $4,409.32
Only null left
Add to Cart
19-153-374
View Documents
PIP™
250331020
Gloss Bronze Anti-fog, Scratch-resistant Clear
Dozen of 12 for $255.11
Case of 12 DZ for $3,061.34
Only null left
Add to Cart
19-153-375
View Documents
PIP™
250331024
Gloss Bronze Anti-fog, Scratch-resistant Brown
Dozen of 12 for $321.12
Case of 12 DZ for $3,853.41
Only null left
Add to Cart
19-153-377
View Documents
PIP™
250331226
Gloss Bronze Anti-fog, Scratch-resistant Blue, Indoor/Outdoor
Dozen of 12 for $350.49
Case of 12 DZ for $4,205.94
Only null left
Add to Cart
19-153-371
View Documents
PIP™
250330010
Gloss Black Anti-reflective, Scratch-resistant Clear N/A N/A N/A
19-153-378
View Documents
PIP™
250331042
Gloss Bronze Scratch-resistant Brown N/A N/A N/A
Description
Description
  • Lightweight, semi-rimless design offers extensive eye coverage and all day comfort
  • Bayonet style temples with non-slip rubber pads provide a secure fit
  • Adjustable rubber nose pads offer a custom, comfortable fit
  • Clear Lens for clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
  • Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
  • Anti-Fog Coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Specifications
Safety Glasses
Reusable
Plastic
Polycarbonate
Polycarbonate
Semi-rimless
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.