Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bovine Serum Albumin Antibody (25G7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP105155 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105155 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105155 Supplier Novus Biologicals Supplier No. NBP105155

Mouse Monoclonal Antibody

Bovine Serum Albumin Monoclonal specifically detects Bovine Serum Albumin in Human, Bovine, Rabbit (Negative) samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Bovine Serum Albumin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 25G7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:1000
Gene Alias albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbols ALB
Host Species Mouse
Immunogen BSA purified from bovine plasma and adsorbed to aluminum hydroxide gel.
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 280717
Test Specificity Reacts strongly with BSA, cross-reacts about 25% with human serum albumin but shows no cross-reaction with rabbit serum albumin.
Target Species Human, Bovine, Rabbit (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.