Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bovine Serum Albumin Antibody (5F9), Novus Biologicals™
SDP

Catalog No. NB600868 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600868 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600868 Supplier Novus Biologicals Supplier No. NB600868
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bovine Serum Albumin Monoclonal specifically detects Bovine Serum Albumin in Bovine samples. It is validated for Dot Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Bovine Serum Albumin
Applications Dot Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5F9
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, ELISA 1:120000, Immunohistochemistry 1:1000-1:5000, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:10-1:500, Dot Blot
Gene Accession No. P02769
Gene Alias albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbols ALB
Host Species Mouse
Immunogen Bovine Serum Albumin Antibody (5F9) was produced by repeated immunizations of balb/c mice with bovine serum albumin. (Uniprot: P02769)
Molecular Weight of Antigen 69.29 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 280717
Test Specificity The antibody recognizes a 66 kDa band corresponding to bovine serum albumin monomer. No cross reactivity is detected against other serum proteins by immunoblot. No reactivity is detected by ELISA against human serum albumin or ovalbumin.
Target Species Bovine
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.