Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BP1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB100481B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100481B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100481B Supplier Novus Biologicals Supplier No. NB100481B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BP1 Polyclonal specifically detects BP1 in Human samples. It is validated for ChIP Assay, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen BP1
Applications ChIP Assay, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q92988
Gene Alias Beta protein 1, BP1distal-less homeo box 7, distal-less homeobox 4, DLX7distal-less homeo box 9, DLX8DLX9, homeobox protein DLX-4, Homeobox protein DLX-7, Homeobox protein DLX-8
Gene Symbols DLX4
Host Species Rabbit
Immunogen A synthetic peptide made specifically to the human BP1 protein sequence (between residues 1-60) [UniProt# Q92988].
Molecular Weight of Antigen 36 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1748
Test Specificity This is specific for human BP1 (DLX4 variant 1). Berg PE, Kirolikar S. DLX4 (distal-less homeobox 4). Atlas Genet Cytogenet Oncol Haematol. January 2011. URL : http://AtlasGeneticsOncology.org/Genes/DLX4ID49827ch17q21.html
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.