Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BP1 Antibody (OTI8A1), PE, Novus Biologicals™
SDP

Catalog No. NB289496 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289496 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289496 Supplier Novus Biologicals Supplier No. NBP270581PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BP1 Monoclonal antibody specifically detects BP1 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen BP1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI8A1
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias Beta protein 1, BP1distal-less homeo box 7, distal-less homeobox 4, DLX7distal-less homeo box 9, DLX8DLX9, homeobox protein DLX-4, Homeobox protein DLX-7, Homeobox protein DLX-8
Host Species Mouse
Immunogen Full length human recombinant protein of human DLX4(NP_612138) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Neuronal Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1748
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.