Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BPGM Antibody (2E11), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB162491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162491 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162491 Supplier Novus Biologicals Supplier No. NBP259493AF594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BPGM Monoclonal antibody specifically detects BPGM in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen BPGM
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 200000000000
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias 2,3-bisphosphoglycerate mutase, bisphosphoglycerate mutase, BPG-dependent PGAM, EC 3.1.3.132,3-bisphosphoglycerate synthase, EC 5.4.2.1, EC 5.4.2.4, erythrocyte 2,3-bisphosphoglycerate mutase2,3-bisphosphoglycerate mutase, erythrocyte
Host Species Mouse
Immunogen Recombinant human BPGM (1-259aa) purified from E. coli.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 669
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.