Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BPI Antibody (971526), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB374610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374610 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374610 Supplier Novus Biologicals Supplier No. FAB74681W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BPI Monoclonal specifically detects BPI in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot.

Specifications

Antigen BPI
Applications Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 971526
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias bactericidal permeability-increasing protein, bactericidal/permeability-increasing protein, CAP 57
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK-293-derived transfected with human BPI, Val32-Lys487, Accession # P17213
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 671
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.