Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BPIFB1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB064466 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB064466 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB064466 Supplier Novus Biologicals Supplier No. NBP300110JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BPIFB1 Polyclonal specifically detects BPIFB1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen BPIFB1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias bA49G10.6, chromosome 20 open reading frame 114, dJ1187J4.1, long palate, lung and nasal epithelium carcinoma associated 1, long palate, lung and nasal epithelium carcinoma-associated protein 1, LPLUNC1, MGC14597, VEMSGP, Von Ebner minor salivary gland protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant BPIFB1 (AAH08429.1; Met1-Gln484).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92747
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.