Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BPNT1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10068164 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10068164 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10068164 Supplier Novus Biologicals Supplier No. NB10068164
Add to Cart
Add to Cart

Goat Polyclonal Antibody

BPNT1 Polyclonal specifically detects BPNT1 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen BPNT1
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:4000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006076.4
Gene Alias 3'(2'), 5'-bisphosphate nucleotidase 1,3'(2')5'-bisphosphate nucleotidase 1, Bisphosphate 3'-nucleotidase 1, BPntase, EC 3.1.3, EC 3.1.3.7, PAP-inositol-1,4-phosphatase, PIP
Gene Symbols BPNT1
Host Species Goat
Immunogen Peptide with sequence C-RNYDYYASRVPESIK corresponding to C-Terminus according to NP_006076.4.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10380
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse and Rat.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.