Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BPOZ Antibody, Novus Biologicals™
SDP

Catalog No. NB100836 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100836 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100836 Supplier Novus Biologicals Supplier No. NB100836
Add to Cart
Add to Cart

Goat Polyclonal Antibody

BPOZ Polyclonal specifically detects BPOZ in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen BPOZ
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Immunohistochemistry 3 - 5 ug/ml, Immunohistochemistry-Paraffin 3 - 5 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_115937.
Gene Alias ankyrin repeat and BTB (POZ) domain containing 1, ankyrin repeat and BTB/POZ domain-containing protein 1, BPOZMGC20585, BTB3, EF1ABP, elongation factor 1A binding protein, Elongation factor 1A-binding protein
Gene Symbols ABTB1
Host Species Goat
Immunogen Peptide with sequence RALEDLLVSIGLDC corresponding to C-Terminus according to NP_115937.1, NP_742024.1, NP_742025.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80325
Test Specificity This antibody is expected to recognize all reported isoforms (NP_115937.1; NP_742024.1; NP_742025.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.