Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brachyury Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB003002 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003002 Supplier Novus Biologicals Supplier No. NBP278841

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Brachyury Polyclonal specifically detects Brachyury in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Brachyury
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunoprecipitation 15 - 20 μL/mg lysate
Formulation Tris-buffered Saline, 0.1% BSA with 0.09% Sodium Azide
Gene Alias brachyury protein, MGC104817, Protein T, SAVA, T brachyury (mouse) homolog, T brachyury homolog, T Brachyury Transcription Factor, T, brachyury homolog (mouse), T-Box Transcription Factor T, TBXT, TF, TFT
Gene Symbols T
Host Species Rabbit
Immunogen The epitope recognized by A305-889A-M maps to a region between residue 350 and 400 of human Brachyury protein using the numbering given in entry O15178.1 (GeneID 6862).
Purification Method Affinity Purified
Quantity 100 μL
Research Discipline Hematopoietic Stem Cell Markers, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6862
Target Species Human
Content And Storage Store at 2-8°C/ 1 year from date of receipt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.