Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bradykinin RB2/BDKRB2 Antibody (471916), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB369202 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369202 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369202 Supplier Novus Biologicals Supplier No. FAB47251Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bradykinin RB2/BDKRB2 Monoclonal specifically detects Bradykinin RB2/BDKRB2 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Bradykinin RB2/BDKRB2
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 471916
Conjugate DyLight 680
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias B2 bradykinin receptor, B2R, BK2, BK-2, BK-2 receptor, BKR2, bradykinin receptor B2, BRB2, DKFZp686O088
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Bradykinin RB2/BDKRB2, Met1-Gln391, Accession # P30411
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 624
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.