Learn More
BRAF Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00607

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine, Dog, Gorilla, Chimpanzee, Northern white-cheeked gibbon, Small-eared galago a
B-raf belongs to the raf/mil family of serine/threonine protein kinases. This protein plays a role in regulating the MAP kinase/ERKs signaling pathway, which affects cell division, differentiation, and secretion. Mutations in the gene are associated with cardiofaciocutaneous syndrome, a disease characterized by heart defects, mental retardation and a distinctive facial appearance. Mutations in this gene have also been associated with various cancers, including non-Hodgkin lymphoma, colorectal cancer, maligt melanoma, thyroid carcinoma, non-small cell lung carcinoma, and adenocarcinoma of lung. A pseudogene, which is located on chromosome X, has been identified for this gene.Specifications
BRAF | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P15056 | |
BRAF | |
Between 25 and 75 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
BRAF | |
B-Raf proto-oncogene serine/threonine-protein kinase; p94; v-Raf murine sarcoma viral oncogene homolog B1; BRAF; BRAF1; RAFB1; | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
673 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.