Learn More
Kodiak™ Bralorne CT CP WP 6 in. Brown Boot
Catalog No. 17100181 Shop All Red Kap Industries Products
Click to view available options
Size:
5
5.5
6
6.5
7
7.5
8
8.5
9
9.5
10
11
Inspired by classic hiking boots and rugged work boots, the look is timeless and premium. Behind the styling, modern work boot technology keeps you safe and dry on the job - with waterproof seam-sealed construction, composite toe and under-foot composite safety plate.
  • Classic Kodiak™ 6 in. boot with bellows leather padded tongue and collar
  • Full grain waterproof leather with waterproof membrane construction
  • Kodiak™ SOS Smell Out Science™ odor control technology
  • Abrasion resistant lining
  • Comfortzone™ footbed with Ortholite™ comfort foam
  • Slip resisting, non-marking rubber outsole
Specifications
Color Brown
Material Leather
Size 10
Gender Women's
Latex Free Yes
Waterproof Yes
Product Type Boot
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.