Learn More
BrandTech™ BRAND™ Dispensette™III, Dispensette™ Organic Remote Dispensing System Parts and Accessories
Catalog No. p-4363608
Click to view available options
Product Type:
Table/Shelf Clamp
This item is not returnable. View return policy
This item is not returnable. View return policy
Allows the Dispensette™ to be mounted on a wall, a ring stand, or on lab furniture.Dispensette™III, Dispensette™ Organic Remote Dispenser Parts and Accessories include threaded adapters, a table/shelf clamp and a support rod for the wall mounting unit.
  • Dispensette™ parts and accessories allow the Dispensette™ to be mounted on a wall, a ring stand, or on lab furniture.
  • Dispenses accurate volumes directly from drums and bulk refills.
  • A filter in the drum adapter minimizes risk of contaminating high-purity reagents.
  • A quick-release connector with integrated valves allows quick changing of the bulk container.
  • The remote dispensing system allows storage of the drum up to 10 meters (30 feet) away from the Dispensette™.
  • Maximum delivery height is approximately 1.2 m
TRUSTED_SUSTAINABILITY
Specifications
Product Type Table/Shelf Clamp
For Use With (Equipment) DispensetteIII, Dispensette Organic
Quantity 1 Ea.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.