Learn More
BRANDTECH™ VITLAB™ Graduated Buckets with Handles
Catalog No. 14379127 Shop All BrandTech Scientific Products
Click to view available options
Material:
HDPE
Includes:
Handle
Catalog No. 14-379-127
Supplier: BRANDTECH™ VITLAB™ 96794
Only null left
Add to Cart
Add to Cart
Durable containers with 1-liter segment divisions and stable handles for easy carrying. Available in white or transparent options, with or without spouts.
Buckets with pour spout
  • Part numbers 96694 and 96794
  • Made of transparent polypropylene
  • Chemical resistant
  • Without lit
Buckets without pour spouts
  • Part numbers 96093 and 96393
  • Made of HDPE
  • Tight fitting, snap-on lid, available separately
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Description 15 L Bucket
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.