Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Self-adhesive Plate Sealing Film

Catalog No. 13878200 Shop All BrandTech Scientific Products
Click to view available options
Material:
Aluminum
Polyester
Polyethylene, Polypropylene
Polypropylene
Rayon
Vinyl
For Use With (Application):
Automation
Cell and tissue culture
Cold storage
ELISA, PCR
ELISA, Real-time PCR
Luminescence measurements
Storage
Storage, fluorescence measurements
No. per Pack:
100 Sheets
50 Sheets

Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.

  • Easily apply to plates and remove without the use of expensive equipment.
  • Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
  • Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
    • Polyethylene
    • Polyester
    • Aluminum
    • Rayon
    • Polypropylene
    • Vinyl
TRUSTED_SUSTAINABILITY

Specifications

Description Highly transparent
Material Polyester
No. per Pack 100 Sheets
For Use With (Application) ELISA, Real-time PCR
Product Type Sealing Film
Temperature Range (Metric) -40°C to +110°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.