Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ BRAND™ 0.2 mL PCR Tubes with Attached Caps, BIO-CERT™ PCR QUALITY

Catalog No. 1388256 Shop All BrandTech Scientific Products
Click to view available options
Color:
Blue
Clear
Green
Rose
Yellow
Closure Type:
Domed Cap
Flat Cap

Thin Wall PCR Tubes with attached caps ensure precise thermal transfer, RNase-/DNase-/DNA-free, and manufactured without lubricants or additives that could contaminate samples.

BRAND ™ Disposable PCR Tubes are designed to deliver exceptional performance for PCR setups, small- and medium-throughput applications, and other workflows requiring precise thermal transfer and high cleanliness. Made from high-quality virgin polypropylene, these tubes ensure reliable results with features that enhance usability and sample integrity. Key features include:

  • BIO-CERT™ PCR QUALITY: Certified free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors, ensuring the highest level of cleanliness for sensitive PCR applications.
  • Superior Heat Transfer: Extra thin-walled tubes optimize thermal transfer during PCR, reducing cycle times and providing consistent performance over a wide temperature range.
  • Reliable Sealing Caps: Tight-sealing caps prevent sample evaporation and loss due to heating, while offering easy-to-open and close functionality.
  • Color Options: Offered in multiple colors for simplified visual sample tracking and organization.
  • High-Quality Construction: Manufactured from high-quality virgin resins without lubricants or chemical additives that could contaminate reagents or samples.
  • Autoclavable: Designed to withstand autoclaving at 121°C (2 bar) in compliance with DIN EN 285.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase and DNA free
Color Green
Volume (Metric) 0.2 mL
Quantity 1000 Pack
For Use With (Equipment) Thermal cyclers for excellent reproducible results in most PCR applications
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap
Autoclavable Autoclavable
Closure Included Yes
Disposable Single-use
Format Tube
Reaction Volume 0.2 mL
Product Type Thin Wall PCR Tube
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.