Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrandTech™ VACUUBRAND™ VHCpro System Accessory - 8-channel Pipet Tip Adapter

Catalog No. 13889230 Shop All BrandTech Scientific Products
Click to view available options
For Use With (Equipment):
VHCPro

VACUUBRAND™ 8-channel pipette tip adapter for VHCpro hand controller

  • Includes tip ejector
TRUSTED_SUSTAINABILITY

Specifications

Product Type 8 Channel Aspirator
Autoclavable Autoclavable
Color Black/White
Channels 8
Material Polypropylene
For Use With (Application) Aspiration from multi-well plates
For Use With (Equipment) VHCPro
Includes 8-channel manifold only
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.