Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRAP Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291567 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291567 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291567 Supplier Novus Biologicals Supplier No. NBP142696F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BRAP Polyclonal antibody specifically detects BRAP in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen BRAP
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias BRAP2RING finger protein 52, BRCA1 associated protein, EC 6.3.2.-, IMPBRCA1-associated protein, Impedes mitogenic signal propagation, Renal carcinoma antigen NY-REN-63, RNF52galectin-2-binding protein
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 560 and 592 of human BRCA1 associated protein using the numbering given in entry NP_006759.3 (GeneID 8315).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8315
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.