Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCA1 Antibody (6B4), Novus Biologicals™
SDP

Catalog No. NB100404 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100404 Supplier Novus Biologicals Supplier No. NB100404
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 6 publications

BRCA1 Monoclonal specifically detects BRCA1 in Human samples. It is validated for Chemotaxis, ChIP Assay, Gene Knock-Out, IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen BRCA1
Applications Chemotaxis, ChIP Assay, Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot
Classification Monoclonal
Clone 6B4
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Knockdown Validated
Gene Alias BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1
Gene Symbols BRCA1
Host Species Mouse
Immunogen BRCA1 protein fragment expressed in E. coli corresponding to amino acids 341-748.
Molecular Weight of Antigen 220 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Tumor Suppressors, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 672
Test Specificity This recognizes BRCA1, a 220-kDa nuclear phosphoprotein, and does not recognize the exon 11 splice variant. Mutations in this tumor suppressor gene greatly increase the risk of breast cancer.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.