Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCA1 Mouse anti-Human, Clone-BRCA1/1398, Novus Biologicals™
SDP

Catalog No. NB254305200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254305200 200 μg
NB254305100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB254305200 Supplier Novus Biologicals Supplier No. NBP254305200UG

Mouse Monoclonal Antibody

IHC-P:Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Tris with 1mM EDTA pH 9.0 for 10-20 min followed by cooling at RT for 20 minutes

Specifications

Antigen BRCA1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone BRCA1/1398
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1
Gene Symbols BRCA1
Host Species Mouse
Immunogen Recombinant human BRCA1 protein fragment (aa445-620) (exact sequence is proprietary)
Molecular Weight of Antigen 220 kDa
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 672
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.