Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCAA1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB345071 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345071 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345071 Supplier Novus Biologicals Supplier No. NB100682APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BRCAA1 Polyclonal specifically detects BRCAA1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen BRCAA1
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias 180 kDa Sin3-associated polypeptide, ARID domain-containing protein 4B, AT rich interactive domain 4B (RBP1- like), AT rich interactive domain 4B (RBP1-like), AT-rich interactive domain-containing protein 4B, BRCAA1DKFZp313M2420, breast cancer-associated antigen 1, Breast cancer-associated antigen BRCAA1, breast carcinoma-associated antigen, Histone deacetylase complex subunit SAP180, Rb-binding protein homolog, RBBP1L1RBP1L1BCAA, RBP1-like protein, retinoblastoma binding protein 1-like 1, Retinoblastoma-binding protein 1-like 1, SAP180MGC163290, SIN3A-associated protein 180, Sin3-associated polypeptide p180
Host Species Rabbit
Immunogen Reacts with residues 1046-1055 [SSKKQKRSHK] of the 136 kDa human, mouse and rat BRCAA1 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 51742
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.