Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCC3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB357277 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357277 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357277 Supplier Novus Biologicals Supplier No. NBP176831MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BRCC3 Polyclonal antibody specifically detects BRCC3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown

Specifications

Antigen BRCC3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias BRCA1/BRCA2-containing complex subunit 3, BRCA1/BRCA2-containing complex subunit 36, BRCA1/BRCA2-containing complex, subunit 3, BRCA1-A complex subunit BRCC36, BRCC36C6.1ACXorf53, BRISC complex subunit BRCC36, c6.1A, chromosome X open reading frame 53, EC 3.1.2.15, EC 3.4.19.-, lys-63-specific deubiquitinase BRCC36
Host Species Rabbit
Immunogen Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human BRCC36. The immunogen is located within amino acids 20 - 70 of BRCC36. Amino Acid Squence: CIGELNDDTRSDSKFAY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79184
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.